Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-BAY-899 | 2988191-40-6 | 99.9% | 50 MG
SDP

Catalog No. 5000338726
Encompass_Preferred

(R)-BAY-899 is the R-enantiomer of BAY-899, an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It exhibits IC50s of 185 nM for human LH and 46 nM for rat LH.

  • Selective luteinizing hormone receptor antagonist
  • Orally active
  • IC50s: 185 nM for hLH, 46 nM for rLH
  • Molecular weight: 459.45
  • Chemical formula: C25H19F2N5O2
  • Appearance: Off-white to light yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 125 mg/mL (272.06 mM)
  • Solubility (in vivo): ≥ 5 mg/mL in various protocols

Catalog No. 50-003-38726 Supplier Medchemexpress LLC Supplier No. HY130248B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-BAY-899 is the R-enantiomer of BAY-899, an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It exhibits IC50s of 185 nM for human LH and 46 nM for rat LH.

  • Selective luteinizing hormone receptor antagonist
  • Orally active
  • IC50s: 185 nM for hLH, 46 nM for rLH
  • Molecular weight: 459.45
  • Chemical formula: C25H19F2N5O2
  • Appearance: Off-white to light yellow solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 125 mg/mL (272.06 mM)
  • Solubility (in vivo): ≥ 5 mg/mL in various protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.