Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Bay-899 | 2988191-40-6 | 99.9% | C25H19F2N5O2 | 10MG
SDP

Catalog No. 5000261243
Encompass_Preferred

(R)-BAY-899 is the R-enantiomer of an orally active, selective luteinizing hormone receptor (LH-R) antagonist supplied for research use. Provided as an off-white to light yellow solid with high reported purity and defined molecular properties, it is intended for biochemical and pharmacological studies including receptor pharmacology and in vitro assays.

  • High purity: 99.9%.
  • Molecular formula: C25H19F2N5O2.
  • Molecular weight: 459.45 g/mol.
  • CAS registry number: 2988191-40-6.
  • Off-white to light yellow solid appearance.
  • Reported activity against LH receptor with IC50s of 185 nM (human) and 46 nM (rat).
  • For research use only; not for human therapeutic use.

Catalog No. 50-002-61243 Supplier Medchemexpress LLC Supplier No. HY130248B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-BAY-899 is the R-enantiomer of an orally active, selective luteinizing hormone receptor (LH-R) antagonist supplied for research use. Provided as an off-white to light yellow solid with high reported purity and defined molecular properties, it is intended for biochemical and pharmacological studies including receptor pharmacology and in vitro assays.

  • High purity: 99.9%.
  • Molecular formula: C25H19F2N5O2.
  • Molecular weight: 459.45 g/mol.
  • CAS registry number: 2988191-40-6.
  • Off-white to light yellow solid appearance.
  • Reported activity against LH receptor with IC50s of 185 nM (human) and 46 nM (rat).
  • For research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.