Learn More
Cayman Chemical (R)-(+)-BAY-K-8644, Purity: ≥95%, CAS: 98791-67-4, 10 mg
SDP
Catalog No. 502969681
Encompass_Preferred
An L-type channel blocker that has negative inotropic and vasodilatory effects at 1 μM
Catalog No. 50-296-9681 Supplier Cayman Chemical Supplier No. 1526310 MG
An L-type channel blocker that has negative inotropic and vasodilatory effects at 1 μM