Learn More
Medchemexpress LLC HY-112316A 10mg Medchemexpress, (R)-BAY1238097 CAS:1564269-85-7 Purity:>98%
SDP
Catalog No. 501969081
Encompass_Preferred
Medchemexpress, HY-112316A 10mg (R)-BAY1238097 CAS:1564269-85-7 (R)-BAY1238097 is the R-isomer with lower activity of BAY1238097. BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9081 Supplier Medchemexpress LLC Supplier No. HY112316A10MG
Medchemexpress, HY-112316A 10mg (R)-BAY1238097 CAS:1564269-85-7 (R)-BAY1238097 is the R-isomer with lower activity of BAY1238097. BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.