Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-Benzyl 2-cyclopropyl-2-hydroxyacetate | 2414393-47-6 | MFCD35244325 | 99.1% | 206.24 | C12H14O3 | 500 MG
SDP

Catalog No. 5000202203
Encompass_Preferred

(R)-Benzyl 2-cyclopropyl-2-hydroxyacetate is a chiral benzyl ester used as a synthetic intermediate in the preparation of pyrrolidines and as a building block for ADC linker/payload chemistry. It is supplied for research use with accompanying COA and SDS documentation.

  • Chiral benzyl ester intermediate for pyrrolidine synthesis and ADC linker construction.
  • CAS number 2414393-47-6.
  • Molecular formula C12H14O3; molecular weight 206.24.
  • Purity listed on product page as approximately 99.1% (product page shows 98.65% and 99.08%).
  • Soluble in DMSO at 200 mg/mL (requires ultrasonic; use newly opened DMSO).
  • Available in 500 MG packaging, with additional sizes including 100 MG, 1 G, 5 G, and 10 G.
  • Storage: pure form -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-02203 Supplier Medchemexpress LLC Supplier No. HY400708500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Benzyl 2-cyclopropyl-2-hydroxyacetate is a chiral benzyl ester used as a synthetic intermediate in the preparation of pyrrolidines and as a building block for ADC linker/payload chemistry. It is supplied for research use with accompanying COA and SDS documentation.

  • Chiral benzyl ester intermediate for pyrrolidine synthesis and ADC linker construction.
  • CAS number 2414393-47-6.
  • Molecular formula C12H14O3; molecular weight 206.24.
  • Purity listed on product page as approximately 99.1% (product page shows 98.65% and 99.08%).
  • Soluble in DMSO at 200 mg/mL (requires ultrasonic; use newly opened DMSO).
  • Available in 500 MG packaging, with additional sizes including 100 MG, 1 G, 5 G, and 10 G.
  • Storage: pure form -20°C for 3 years or 4°C for 2 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.