Learn More
Medchemexpress LLC (R)-benzyl mandelate | 97415-09-3 | MFCD00674031 | ≥99.0% | 242.27 g/mol | C15H14O3 | 50 G
SDP
Catalog No. 5000301321
Encompass_Preferred
(R)-Benzyl mandelate is the R-enantiomer of benzyl mandelate (D-(-)-mandelic acid benzyl ester), a chiral synthetic intermediate used in pharmaceutical and organic synthesis. It is a white solid with molecular formula C15H14O3 and molecular weight 242.27 g/mol (CAS 97415-09-3). Recommended storage: keep container tightly sealed at room temperature; for long-term storage in solvent, keep at -20 °C to -80 °C.
Catalog No. 50-003-01321 Supplier Medchemexpress LLC Supplier No. HYW02065550G
(R)-Benzyl mandelate is the R-enantiomer of benzyl mandelate (D-(-)-mandelic acid benzyl ester), a chiral synthetic intermediate used in pharmaceutical and organic synthesis. It is a white solid with molecular formula C15H14O3 and molecular weight 242.27 g/mol (CAS 97415-09-3). Recommended storage: keep container tightly sealed at room temperature; for long-term storage in solvent, keep at -20 °C to -80 °C.