Learn More
TARGETMOL CHEMICALS INC R-BI-2852 5MG
SDP
Catalog No. 502579241
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (R)-BI-2852 can be used as (R)-5-isoindolin-1-one is used as an inhibitor of the RAS family of proteins and as a compound for the treatment and prevention of tumours. (R)-BI-2852 has Antiproliferative and antitumour agent activity. purity: 97%
Catalog No. 50-257-9241 Supplier TARGETMOL CHEMICALS INC Supplier No. T720845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (R)-BI-2852 can be used as (R)-5-isoindolin-1-one is used as an inhibitor of the RAS family of proteins and as a compound for the treatment and prevention of tumours. (R)-BI-2852 has Antiproliferative and antitumour agent activity. purity: 97%