Learn More
Medchemexpress LLC HY-19778 5mg Medchemexpress, (R)-BPO-27 CAS:1415390-47-4 Purity:>98%
SDP
Catalog No. 502082184
Encompass_Preferred
Medchemexpress, HY-19778 5mg (R)-BPO-27 CAS:1415390-47-4 (R)-BPO-27, the R enantiomer of BPO-27, is a potent, orally active and ATP-competitive CFTR inhibitor with an IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2184 Supplier Medchemexpress LLC Supplier No. HY197785MG
Medchemexpress, HY-19778 5mg (R)-BPO-27 CAS:1415390-47-4 (R)-BPO-27, the R enantiomer of BPO-27, is a potent, orally active and ATP-competitive CFTR inhibitor with an IC50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.