Learn More
Cayman Chemical (R)-Bromoenol lactone, 478288-90-3, 1 mg
SDP
Supplier:  Cayman Chemical 100068001
Encompass_Preferred
The PLAs are an extensive family of lipid hydrolases that function in cell signaling, digestion, membrane remodeling, and as venom components.{6940} The iPLA2 are a PLA2 subfamily closely associated with the release of arachidonic acid in response to physiologic stimuli. (R)-BEL is an irreversible, chiral, mechanism-based inhibitor of iPLA2γ. Unlike (S)-BEL, (R)-BEL does not inhibit iPLA2β except at high doses of 20-30 µM.{12978} (R)-BEL inhibits human recombinant iPLA2γ with an IC50 of approximately 0.6 µM.
Catalog No. NC1379738