Learn More
Electron Microscopy Sciences R-Buffer Bg(2x), 250 ml for use in Retriever Thermal Slide Processor 250 ML
SDP
Catalog No. 5031182
Encompass_Preferred
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.
Catalog No. 50-311-82 Supplier Electron Microscopy Sciences Supplier No. 6270711
Used with the Retriever benchtop thermal slide processer that is used on formalin-fixed, paraffin embedded tissues prior to immunostaining.