Learn More
Medchemexpress LLC (R)-Carnitine-d3 (hydrochloride); Levocarnitine-d3 (hydrochloride) | 350818-62-1 | 99.9% | 5 MG
SDP
Catalog No. 5000413039
Encompass_Preferred
L-Carnitine-d3 (hydrochloride) is a deuterium-labeled form of L-Carnitine hydrochloride. It is intended for research use only.
  • Deuterium labeled: This compound is the deuterium labeled L-Carnitine hydrochloride.
  • Research use: Primarily used for research purposes.
  • Tracer: Can be utilized as a tracer in various studies.
  • Internal standard: Suitable as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Pharmacokinetic studies: Deuteration can influence the pharmacokinetic and metabolic profiles of drugs, making this compound valuable for such investigations.
Catalog No. 50-004-13039 Supplier Medchemexpress LLC Supplier No. HYB1453S5MG
L-Carnitine-d3 (hydrochloride) is a deuterium-labeled form of L-Carnitine hydrochloride. It is intended for research use only.
  • Deuterium labeled: This compound is the deuterium labeled L-Carnitine hydrochloride.
  • Research use: Primarily used for research purposes.
  • Tracer: Can be utilized as a tracer in various studies.
  • Internal standard: Suitable as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Pharmacokinetic studies: Deuteration can influence the pharmacokinetic and metabolic profiles of drugs, making this compound valuable for such investigations.