Learn More
Medchemexpress LLC HY-B0006C 5mg Medchemexpress, (R)-Carvedilol CAS:95093-99-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0006C5MG
Encompass_Preferred
Medchemexpress, HY-B0006C 5mg (R)-Carvedilol CAS:95093-99-5 (R)-Carvedilol ((R)-BM 14190), the R-enantiomer of Carvedilol, is a non-selective β/α-1 blocker. (R)-Carvedilol exerts protection against the vascular or cardiac toxicity of Doxorubicin (DOX). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2343