Learn More
Medchemexpress LLC (R)-carvedilol | 95093-99-5 | MFCD00869663 | 98.8% | 406.47 g/mol | C24H26N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0006C10MG
Encompass_Preferred
(R)-Carvedilol is the R-enantiomer of carvedilol provided for research use. It functions primarily as an α-receptor blocker with minimal β-blocking activity and has reported effects on spontaneous Ca2+ waves and stress-induced ventricular tachycardia; formulating examples and solubility data are available for in vitro and in vivo use.
  • Orally active R-enantiomer with selective α-receptor blocking activity.
  • High purity suitable for analytical and in vitro studies.
  • Soluble in DMSO at high concentrations; documented in vivo formulation protocols.
  • Known chemical formula and molecular weight for formulation and dosing calculations.
  • Available in small pack sizes for stereochemistry and pharmacology research.
Catalog No. 50-002-99245