Learn More
Strem, An Ascensus Company CAS 112022-83-0. 10ml. (R)-Tetrahydro-1-methyl-33-diphenyl-1H3H-pyrrolo[12-c][132]oxazaborole 0.9-1.1M in toluene [(R)-Methyloxazaborolidine] (R)-CBS Catalyst. MFCD00078440
SDP
Catalog No. 502110680
Encompass
CAS 112022-83-0. 10ml. (R)-Tetrahydro-1-methyl-33-diphenyl-1H3H-pyrrolo[12-c][132]oxazaborole 0.9-1.1M in toluene [(R)-Methyloxazaborolidine] (R)-CBS Catalyst. MFCD00078440. Molecular Weight 277.17. Molecular Formula C18H20BNO. Color/form liq. Strem 05-1000. http//www.strem.com/catalog/v/05-1000/
Catalog No. 50-211-0680 Supplier Strem, An Ascensus Company Supplier No. 05100010ML
CAS 112022-83-0. 10ml. (R)-Tetrahydro-1-methyl-33-diphenyl-1H3H-pyrrolo[12-c][132]oxazaborole 0.9-1.1M in toluene [(R)-Methyloxazaborolidine] (R)-CBS Catalyst. MFCD00078440. Molecular Weight 277.17. Molecular Formula C18H20BNO. Color/form liq. Strem 05-1000. http//www.strem.com/catalog/v/05-1000/