Learn More
Cayman Chemical R-CCG-1423 100ug
SDP
Catalog No. 502983406
Encompass_Preferred
A stereoisomer of CCG-1423 that less potently inhibits MRTF-A-dependent cellular events, including SRF-mediated gene expression and cell migration
Catalog No. 50-298-3406 Supplier Cayman Chemical Supplier No. 20106100 UG
A stereoisomer of CCG-1423 that less potently inhibits MRTF-A-dependent cellular events, including SRF-mediated gene expression and cell migration