Learn More
Cayman Chemical (R)-Cetirizine (hydrochloride), Purity: ≥98%, CAS: 130018-87-0, 50 mg
SDP
Catalog No. 502934611
Encompass_Preferred
A selective histamine H1 receptor antagonist; the (R)-enantiomer of cetirizine; binds to the H1 receptor with higher affinity than cetirizine (Kis = 3 and 6 nM, respectively) and is highly selective for H1 over M1-M5 receptors; decreases production of RANTES and eotaxin following antigen stimulation in mouse eosinophils in vitro (MEC = 0.05 µM); dose-dependently decreases histamine-induced nasal rubbing and sneezing in mice (0.01-1% solution)
Catalog No. 50-293-4611 Supplier Cayman Chemical Supplier No. 2399250 MG
A selective histamine H1 receptor antagonist; the (R)-enantiomer of cetirizine; binds to the H1 receptor with higher affinity than cetirizine (Kis = 3 and 6 nM, respectively) and is highly selective for H1 over M1-M5 receptors; decreases production of RANTES and eotaxin following antigen stimulation in mouse eosinophils in vitro (MEC = 0.05 µM); dose-dependently decreases histamine-induced nasal rubbing and sneezing in mice (0.01-1% solution)