Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-citronellol | 1117-61-9 | MFCD00063215 | 99.8% | 156.27 g/mol | C10H20O | 100MG
SDP

Catalog No. 5000239429
Encompass_Preferred

(R)-Citronellol is the R-enantiomer of the monoterpenoid alcohol citronellol, used as a chiral probe and as a fragrance/flavor building block in laboratory research. It is provided as a high-purity liquid suitable for analytical and preparative work.

  • Chiral monoterpenoid alcohol; CAS 1117-61-9.
  • Chemical formula C10H20O; molecular weight 156.27 g/mol.
  • Purity 99.8%.
  • Physical form liquid; colorless to light yellow; density 0.857 g/cm3.
  • Storage: 4°C sealed; in solvent keep at -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 50 mg, 100 mg, 200 mg, 500 mg; also available as a 10 mM solution in DMSO (1 mL).

Catalog No. 50-002-39429 Supplier Medchemexpress LLC Supplier No. HY124257100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Citronellol is the R-enantiomer of the monoterpenoid alcohol citronellol, used as a chiral probe and as a fragrance/flavor building block in laboratory research. It is provided as a high-purity liquid suitable for analytical and preparative work.

  • Chiral monoterpenoid alcohol; CAS 1117-61-9.
  • Chemical formula C10H20O; molecular weight 156.27 g/mol.
  • Purity 99.8%.
  • Physical form liquid; colorless to light yellow; density 0.857 g/cm3.
  • Storage: 4°C sealed; in solvent keep at -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 50 mg, 100 mg, 200 mg, 500 mg; also available as a 10 mM solution in DMSO (1 mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.