Learn More
Abcam (R)-CPPene (SDZ EAA 494), competitive NMDA antagonist, 10MG
Catalog No. 502707773
Encompass_Preferred
MW 250.19 Da, Purity >98%. Potent competitive NMDA antagonist. Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7773 Supplier Abcam Supplier No. AB12023210MG
MW 250.19 Da, Purity >98%. Potent competitive NMDA antagonist. Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement