Learn More
AdipoGen (R)-CR8 (1 mg)
SDP
Supplier:  AdipoGen AGCR10039M001
Encompass_Preferred
Chemical. CAS: 294646-77-8. Formula: C24H29N7O. MW: 431.5. Potent and selective cyclin-dependent kinase (CDK) 1, 2, 5, 7 and 9 inhibitor. Apoptosis inducer. Potent glycogen synthase kinase (GSK-3alpha/beta) inhibitor.
Catalog No. 50-205-3795