Learn More
AdipoGen (R)-CR8 (50 mg)
SDP
Supplier:  AdipoGen AGMRC0003M050
Encompass_Preferred
Chemical. CAS: 294646-77-8. Formula: C24H29N7O. MW: 431.5. Potent and selective inhibitor of cyclin dependent kinases CDK1, CDK2, CDK5, CDK7 and CDK9. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Casein kinase 1 (CK1delta/epsilon) and glycogen synthase kinase (GSK-3alpha/beta) inhibitor. Potential anti-inflammatory compound. Potential antidiabetic compound.
Catalog No. 50-205-4224