Learn More
Medchemexpress LLC HY-18340A 5mg Medchemexpress, (R)​-​CR8 (trihydrochloride) CAS:1786438-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18340A5MG
Encompass_Preferred
Medchemexpress, HY-18340A 5mg (R)​-​CR8 (triHCl) CAS:1786438-30-9 (R)-CR8 trihydrochloride (CR8 trihydrochloride), a second-generation analog of Roscovitine, is a potent CDK1/2/5/7/9 inhibitor. (R)-CR8 trihydrochloride inhibits CDK1/cyclin B (IC50=0.09 μM), CDK2/cyclin A (0.072 μM), CDK2/cyclin E (0.041 μM), CDK5/p25 (0.11 μM), CDK7/cyclin H (1.1 μM), CDK9/cyclin T (0.18 μM) and CK1δ/ε (0.4 μM). (R)-CR8 trihydrochloride induces apoptosis and has neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0007