Learn More
Apexbio Technology LLC (R)-Crizotinib, 10mg. Cas: 877399-52-5 MFCD: MFCD12407409. C-MET/ALK inhibitor,potent and ATP-competitve
SDP
Supplier:  Apexbio Technology LLC A302010
Encompass_Preferred
Crizotinib is a potent, ATP-competitive, small-molecule and orally available inhibitor of c-Met kinase with a Ki valueof 4 nmol/L[1].Crizotinib has shown to inhibitwild-type c-Met phosphorylation with a mean IC50 value of 11 nmol/L in multiple human endothelial and carcinoma cell lines. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4782