Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Amphibian TGF-beta 5 Protein

Catalog No. p-4718853
Change view
Click to view available options
Quantity:
10 μg
2 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
245B5002 2 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
245B5002CF 2 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
245B5010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
245B5010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 245B5002 Supplier R&D Systems™ Supplier No. 245B5002
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Amphibian TGF-beta 5 Protein is derived from Sf 21 (baculovirus). The Recombinant Amphibian TGF-beta 5 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Gene ID (Entrez) 397778
Molecular Weight (g/mol) 12.6 kDa
Name TGF-beta 5
Quantity 2 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived amphibian TGF-beta 5 protein Gly271-Ser382 & Gln274-Ser382
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstituted at 20 μg/mL in sterile 4 mM HCl containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.