Learn More
R&D Systems™ Boc-VPR-AMC Fluorogenic Peptide Substrate

Description
- Substrate for Coagulation Factor II, Coagulation Factor VII, Kallikrein 4, Kallikrein 5, Kallikrein 8, Kallikrein 13, Kallikrein 14, and MASP3
- Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
- A fluorescence plate reader with excitation at 380nmand emission at 460nmis recommended for the measurement of the enzymatic activity
- Purity >95% based on TLC
Specifications
Specifications
| Quantity | 20 mg |
| Product Type | Boc-Val-Pro-Arg-AMC |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.