Learn More
R&D Systems™ DLDVPIPGRFDRRVS(PO3)VAAE Ser/Thr Phosphatase Substrate I

Description
- Synthetic phosphopeptide corresponding to amino acids 81 to 99 of Protein Kinase A Regulatory Subunit 2-alpha
- Enzymatic cleavage of this substrate liberates inorganic phosphate, which can be measured in dye binding assays, such as the malachite green method
- Purity 95% based on HPLC analysis
Specifications
Specifications
| Form | Lyophilized |
| Quantity | 1 mg |
| Format | Lyophilized from a stock solution in distilled water. |
| Molecular Weight (g/mol) | Theoretical Molecular weight: 2192 Da |
| Product Type | Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser(PO3)-Val-Ala-Ala-Glu |
| Storage Requirements | No Stability Data Storage is set to N/A for ProdCode |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.