Learn More
R&D Systems™ H-Met-Gly-Pro-AMC Fluorogenic Peptide Substrate
SDP
Catalog No. ES017
Change view
Click to view available options
Quantity:
5 mg
Catalog No. Quantity
ES017 5 mg
1 options
This item is not returnable. View return policy
Catalog No. ES017 Supplier R&D Systems™ Supplier No. ES017
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Substrate for Methionine Aminopeptidase 1, 1D and 2
  • Hydrolysis of Pro-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Excellent substrate for methionine aminopeptidases; after the enzymatic removal of the Met residue, the substrate can be cleaved by DPPIV/CD26-like enzymes
  • Purity >97% by TLC
Specifications
Quantity 5 mg
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.