Learn More
R&D Systems™ HT 8-oxo-dG ELISA II Kit
Description
Includes
Plate; 20uM 8-oxo-dG Standard, 20uL; Anti-8-oxo-dG, 20uL; Goat anti-Mouse IgG-HRP, 20uL; 100X Cations, 500uL; Dnase I 5 units/uL, 40uL; Alkaline Phosphatase 1 unit/uL, 40uL; TACS-Sapphire, 10mL; Quantities doubled for Mfr. No. 4380-192-K.
8-oxo-dG is a frequently used biomarker of oxidative DNA damage and oxidative stress. Reactive oxygen and nitrogen species generated by normal metabolic processes and a variety of environmental factors cause the hydroxylation of guanosine at the N7-C8 bon
Provides cations, DNase I, and alkaline phosphatase for prepping DNA extracted from cultured cells, adherent cells, and tissue for use in the assay
This ELISA uses TACS-Sapphire™ as the colorimetric, non-radioactive substrate for HRP
A
Specifications
Specifications
| Includes | 20μM 8-oxo-dG Standard, Anti-8-oxo-dG, 20μL, Goat anti-Mouse IgG-HRP, 100X Cations, Dnase I, Alkaline Phosphatase, TACS-Sapphire |
| Quantity | 1 x 96 Tests |
| Content And Storage | The product requires storage at both -20°C to -70°C and 2°C to 8°C. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Product Type | HT 8-oxo-dG ELISA II Kit |