Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Airway Trypsin-like Protease/HAT, CF

Catalog No. 2695SE010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2695SE010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2695SE010 Supplier R&D Systems™ Supplier No. 2695SE010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, t-butoxycarbonyl-Phe-Ser-Arg-7-amino-4-methyl coumarin (Boc-FSR-AMC). The specific activity is >50,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O60235
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 9407
Molecular Weight (g/mol) M.W. (Observed): 27 kDa and 18 kDa, reducing conditions; M.W. (theoretical): 25 kDa & 17 kDa
Name Airway Trypsin-like Protease/HAT
Quantity 10 μg
Source Mouse myeloma cell line, NS0-derived human Airway Trypsin-like Protease/HAT protein Ala42-Ile418 with an N-terminal 6-His tag The protein was purified, activated and further purified.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Active
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol TMPRSS11D
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile 25 mM Tris, 150 mM NaCl and 0.05% Brij-35, pH 7.5.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Airway Trypsin-like Protease/HAT/TMPRSS11D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.