Learn More
R&D Systems™ Human alpha 2-Macroglobulin Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and Glycerol. |
| Gene ID (Entrez) | 2 |
| Name | alpha 2-Macroglobulin |
| Quantity | 50 μg |
| Source | Human plasma-derived alpha 2-Macroglobulin protein The human plasma used for the isolation of this product was certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked homo-oligomer |
| Conjugate | Unconjugated |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.