Learn More
R&D Systems™ Human CD40/TNFRSF5 DuoSet ELISA

Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Conjugate | Biotin |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 958 |
| Gene Symbol | CD40 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 |
| Target | CD40/TNFRSF5 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.