Learn More
R&D Systems™ Human CD47 DuoSet ELISA

Description
Includes
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
Specifications
Specifications
| Assay Range | 125.00 to 8000 pg/mL |
| Conjugate | Biotin |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 961 |
| Gene Symbol | CD47 |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen |
| Target | CD47 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.