Learn More
R&D Systems™ Human CXCL10/IP-10 EliSPOT™ Development Module

Description
Includes
Human CXCL10/IP-10/CRG-2 Capture Antibody, Human CXCL10/IP-10/CRG-2 Biotinylated-conjugated Detection Antibody
Contains the basic components required to develop an ELISpot (quantitative sandwich ELISA) assay
Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
Generic development protocols provide directi
Specifications
Specifications
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.