Learn More
R&D Systems™ Human CXCL9/MIG ELISpot Development Module
SDP
Catalog No. SEL392
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SEL392 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SEL392 Supplier R&D Systems™ Supplier No. SEL392
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Sufficient for at least five 96-well plates
Product Features:
  • An economical alternative to ELISpot Kits
  • Optimized capture and detection antibody pairings and recommended concentrations save lengthy development time
  • Generic development protocols provide direction to start an optimization protocol
  • Customize the assay to your specific needs
Module content:
  • Capture Antibody
  • Biotinylated-conjugated Detection Antibody
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.