Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human GDF-15 Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Manufacturer:  R&D Systems™ 957GD025/CF

 View more versions of this product

Catalog No. 957GD025CF


Add to cart
This item is not returnable. View return policy

Description

Description

The Recombinant Human GDF-15 Protein is derived from CHO.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Predicted MW: 13kDa (monomer) Observed MW: 14kDa, reducing conditions
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Source: Chinese Hamster Ovary cell line, CHO-derived Ala197-Ile308, with an N-terminal 6-His tag; N Terminal Sequence: His & Asn199
Disulfide-linked homodimer
Yes
9518
GDF-15
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Reconstitute at 100-300μg/mL in sterile 4mM HCI.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit