Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human His6-UbcH10/UBE2C Protein, CF
Description
Recombinant Human His6-UbcH10/UBE2C is a member of the Ubiquitin-conjugating (E2) enzyme family that receives Ubiquitin from a Ubiquitin-activating (E1) enzyme and subsequently interacts with a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human His6-UbcH10/UBE2C concentration of 0.1-1 μM.
Specifications
Specifications
| Chemical Name or Material | His6 UbcH10/UBE2C |
| For Use With (Application) | Enzyme Activity |
| Quantity | 100 μg |
| Formulation | X mg/mL (X μM) in 50 mM HEPES pH 8.0, 200 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP |
| Percent Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?