Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human His6 Ubiquitin Mutant with K29 only Recombinant Protein

Catalog No. UMHK29001M
Change view
Click to view available options
:
1mg; Carrier Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
UMHK29001M 1mg; Carrier Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UMHK29001M Supplier R&D Systems™ Supplier No. UMHK29001M
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The lysine residue utilized for Ubiquitin chain formation is functionally important. Single lysine Ubiquitin mutants are ideal for investigating biological processes involving specific Ubiquitin chain linkage. Recombinant Human His6-Ubiquitin K29-only allows for the formation of only K29-linked Ubiquitin chains. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human His6-Ubiquitin K29-only concentration of 10-50 μM.

Specifications

Accession Number P0CG47
Formulation No Formulation Info in Bulk Inventory
Gene ID (Entrez) 7314
Molecular Weight (g/mol) 9.8 kDa
Name His6 Ubiquitin Mutant with K29 only
Quantity 1mg
Source E. coli-derived human Ubiquitin protein Contains a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute in deionized water or aqueous buffer.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.