Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human HVEM/TNFRSF14 Fc Chimera Protein, CF

Catalog No. 356HV100CF
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
356HV100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 356HV100CF Supplier R&D Systems™ Supplier No. 356HV100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit TNF-beta-mediated cytotoxicity using L-929 mouse fibroblast cells. The ED50 for this effect is 2.5-10 μg/mL in the presence of 50 pg/mL of Recombinant Human TNF-beta/TNFSF1 (Catalog # 211-TB ).

Specifications

Accession Number Q92956.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8764
Molecular Weight (g/mol) M.W. (Observed): 60 kDa, reducing conditions; M.W. (theoretical): 45 kDa (monomer)
Name HVEM/TNFRSF14 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human HVEM/TNFRSF14 protein Human HVEM Pro37-Val202 (Ser108Thr; Ala140Arg) (N-terminus) Accession # Q92956.3 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol TNFRSF14
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein HVEM/TNFRSF14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.