Learn More
R&D Systems™ Human IL-9 ELISpot Development Module
SDP
Catalog No. SEL209
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SEL209 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SEL209 Supplier R&D Systems™ Supplier No. SEL209
Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Sufficient for at least five 96-well plates
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.