Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse Ephrin Sampler Pack Protein Sampler Pack
SDP

Catalog No. SMPK3 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SMPK3 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SMPK3 Supplier R&D Systems™ Supplier No. SMPK3

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Eph receptors represent the largest family of receptor tyrosine kinases (RTK) in the human genome. Their ligands, the ephrins, are also cell surface bound proteins, thereby requiring cell-cell interaction for binding and signaling. The ephrin ligands are divided into two categories: the A class are tethered to the membrane by a GPI anchor, and the B class are transmembrane proteins. Likewise, the receptors are also divided into two categories based on sequence similarity and ligand specificity. Ephrin-A ligands bind EphA receptors and ephrin-B ligands bind EphB receptors, with the exception of EphA4 which binds both A and B-type ligands. Eph/ephrin signaling is involved in vascular development, tissue-border formation, brain boundaries, cell migration, axon guidance, and synaptic plasticity. Eph/ephrin signaling activation generally results in repulsion of cells and often converges on the regulation of the cytoskeleton. Like most RTKs, ligand binding induces forward signaling, where a response occurs in the Eph receptor-containing cell. However, in some circumstances, this family can also exhibit reverse signaling where ephrins act as receptors responding to Eph signals, and ephrins signal back into their host cell.

Specifications

Formulation Lyophilized
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Pack
Label or Dye Unconjugated
Species Human/Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.