Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Pro-Collagen II Protein
Click to view available options
:
20ug; Carrier Free
Description
The Recombinant Human Pro-Collagen II Protein is derived from CHO. The Recombinant Human Pro-Collagen II Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in Tris and NaCl. |
Gene ID (Entrez) | 1280 |
Molecular Weight (g/mol) | 78 kDa |
Name | Pro Collagen II |
Quantity | 20μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Collagen II protein Gln26-Arg266 & Gly957-Leu1487,with an N-terminal 5-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction