Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus
GSA_VA

Catalog No. p-4720779
Change view
Click to view available options
Quantity:
100 μg
25 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
1845SH025 25 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
1845SH100 100 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
1845SH025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
1845SH100CF 100 μg Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 1845SH025 Supplier R&D Systems™ Supplier No. 1845SH025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus is derived from E. coli. The Recombinant Human Sonic Hedgehog/Shh (C24II) N-Terminus has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number NP_000184.1
Formulation Lyophilized from a 0.2μm filtered solution in PBS and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 6469
Molecular Weight (g/mol) 20 kDa
Name Sonic Hedgehog/Shh (C24II), N-Terminus
Quantity 25 μg
Source E. coli-derived human Sonic Hedgehog/Shh protein Cys24-Gly197 (Cys24Ile-Ile),with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100-200 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin, and allow up to 24 hours at 2 to 8° C for complete reconstitution.
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.