Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165b Protein

Catalog No. 3045VE025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
3045VE025 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
3045VE025CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3045VE025 Supplier R&D Systems™ Supplier No. 3045VE025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. Serially diluted Recombinant Human (rh) VEGF165b is added to a Recombinant Human VEGF R2/KDR/Flk-1 Fc Chimera (Catalog # 357-KD ) coated plate (2 μg/mL, 100 μL/well). Bound rhVEGF165b is detected by a human VEGF165b specific antibody (Catalog # MAB3045 ). The concentration of rhVEGF165b that produces half-maximum response is approximately 0.12-0.6 nM.

Specifications

Accession Number AAL27435
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 7422
Molecular Weight (g/mol) M.W. (Observed): 19-24 kDa, reducing conditions; M.W. (theoretical): 19.1 kDa (monomer)
Name VEGF 165b
Quantity 25 μg
Source Spodoptera frugiperda, Sf 21 (stably transfected)-derived human VEGF protein Ala27-Asp191
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol VEGFA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein VEGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.