Learn More
Hyaluronan (High Molecular Weight), R&D Systems™

Specifications
Specifications
| Chemical Name or Material | Hyaluronan |
| Color | White |
| Physical Form | White to off-white odorless powder |
| Quantity | 100 mg |
| Odor | Odorless |
| Synonym | Hyaluronic acid (HA), Sodium hyaluronate |
| Formula Weight | Predicted MW: >950kDa Observed MW: 1350kDa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.