Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Mca-PLAQAV-Dpa-RSSSR-NH2 Fluorogenic Peptide Substrate
SDP

Catalog No. ES003
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES003 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ES003 Supplier R&D Systems™ Supplier No. ES003

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications: Bioactivity

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Also an excellent substrate for ADAM8

Specifications

Form Liquid
For Use With (Application) Bioactivity
Quantity 1 mg
Format Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 8.0mg/mL or 4 mM.
Molecular Weight (g/mol) Predicted MW: 1638 Da
Product Type Source: Synthetic
Storage Requirements Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.