Learn More
R&D Systems™ Mca-SEVNLDAEFRK(Dnp)RR-NH2 Fluorogenic Peptide Substrate

Description
- Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
- It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
- Purity 93.8% by HPLC
- Contains the Swedish mutation of the amyloid precursor protein (APP) beta-secretase cleavage site
- A K-R-R sequence is added for easy solubilization
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Bioactivity |
| Quantity | 1 mg |
| Format | Supplied as a stock solution in dimethyl sulfoxide (DMSO) (100μL) at a concentration of 10.0mg/mL or 4.1 mM.. |
| Molecular Weight (g/mol) | Predicted MW: 2001 Da |
| Product Type | Source: Synthetic |
| Storage Requirements | Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.