Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse BACE-2 Protein, CF

Catalog No. 2977AS020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2977AS020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2977AS020 Supplier R&D Systems™ Supplier No. 2977AS020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (Catalog # ES010 ). The specific activity is >50 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9JL18
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 25825
Molecular Weight (g/mol) M.W. (Observed): 55-76 kDa, reducing conditions; M.W. (theoretical): 48 kDa
Name BACE-2
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived mouse BACE-2 protein Phe29-Pro462 (Tyr357His) with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Bace2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile, deionized water.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein BACE-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.