Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Cathepsin C/DPPI Protein
Click to view available options
Quantity:
10 μg
Description
The Recombinant Mouse Cathepsin C/DPPI Protein is derived from NS0. The Recombinant Mouse Cathepsin C/DPPI Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 13032 |
| Molecular Weight (g/mol) | 51 kDa |
| Name | Cathepsin C/DPPI |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Cathepsin C/DPPI protein Asp25-Leu462,with a C-terminal 10-His tag Accession &Num; P97821 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Pro form |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction