Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Clusterin Protein

Catalog No. 2747HS050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy

Catalog No. 2747HS050

Supplier: R&D Systems™ 2747HS050

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Clusterin Protein is derived from NS0. The Recombinant Mouse Clusterin Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 12759
Molecular Weight (g/mol) Predicted MW: 23.9kDa (beta subunit) and 26.3kDa (alpha subunit) Observed MW: 37kDa (beta subunit) and 44kDa (subunit), reducing conditions
Name Clusterin
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Clusterin protein Glu22-Arg226 (beta) & Ser227-Glu448 (alpha),with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked heterodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.