Learn More
R&D Systems™ Recombinant Mouse IFN-alpha 1 Protein
Specifications
Specifications
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |