Learn More
R&D Systems™ Mouse IL-12/IL-23 p40 ELISpot Development Module
Catalog No. SEL499
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
SEL499 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. SEL499 Supplier R&D Systems™ Supplier No. SEL499
This item is not returnable. View return policy
Sufficient for at least five 96-well plates
Specifications
Quantity 1 Kit
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.